Skip to Content
Merck
All Photos(1)

Key Documents

C66056

Sigma-Aldrich

4-(4-Chlorophenyl)-4-hydroxypiperidine

99%

Synonym(s):

Haloperidol metabolite I, 4-(4-Chlorophenyl)-4-hydroxypiperidine, 4-(4-Chlorophenyl)-4-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClNO
CAS Number:
Molecular Weight:
211.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

137-140 °C (lit.)

SMILES string

OC1(CCNCC1)c2ccc(Cl)cc2

InChI

1S/C11H14ClNO/c12-10-3-1-9(2-4-10)11(14)5-7-13-8-6-11/h1-4,13-14H,5-8H2

InChI key

LZAYOZUFUAMFLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Fang et al.
Journal of chromatography. B, Biomedical applications, 682(2), 283-288 (1996-07-12)
An electron-capture gas chromatographic procedure was developed for the analysis of 4-(4-chlorophenyl)-4-hydroxypiperidine (CPHP), a metabolite of haloperidol. The assay involved basic extraction of this metabolite from the biological samples, followed by back-extraction with HCl. After basification of the acid phase
Ilse Weuts et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 25(4-5), 387-393 (2005-05-17)
The purpose of the study was to investigate the suitability of polyacrylic acid (PAA) as a carrier in solid dispersions, with the aim to delay crystallization of basic drugs and improve their dissolution behaviour. The physicochemical properties were investigated in
A B Skorniakova et al.
Sudebno-meditsinskaia ekspertiza, 52(1), 45-48 (2009-04-18)
Haloperidol is extensively used in current psychiatric practice for the treatment of various psychotic disorders. However, this substance is known to be toxic and sometimes cause poisoning despite its generally positive therapeutic effect. Moreover, some of its metabolites also possess
Yasuhiko Higashi et al.
Biomedical chromatography : BMC, 20(9), 964-970 (2006-03-01)
4-(4-Chlorophenyl)-4-hydroxypiperidine (CPHP), one of the metabolites of haloperidol, is considered to exhibit brain toxicity. CPHP concentrations in plasma and tissue homogenates (each 200 microL) from rats were analyzed by HPLC fluorescence detection after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F). After basic
Tetsuya Arinobu et al.
Journal of the American Society for Mass Spectrometry, 13(3), 204-208 (2002-03-23)
Sonic spray ionization (SSI) was compared with atmospheric pressure chemical ionization (APCI) as an interface of high-performance liquid chromatography (HPLC)-mass spectrometry (MS) for sensitive analyses of a neuroleptic drug, haloperidol and its two metabolites, such as reduced haloperidol and 4-(4-chlorophenyl)-4-hydroxypiperidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service