C42807
1-(2-Chloroethyl)pyrrolidine hydrochloride
98%
Synonym(s):
2-Pyrrolidinoethyl chloride hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H12ClN · HCl
CAS Number:
Molecular Weight:
170.08
Beilstein:
3677061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
crystals
mp
167-170 °C (lit.)
SMILES string
Cl[H].ClCCN1CCCC1
InChI
1S/C6H12ClN.ClH/c7-3-6-8-4-1-2-5-8;/h1-6H2;1H
InChI key
FSNGFFWICFYWQC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service