Skip to Content
Merck
All Photos(1)

Key Documents

C352

Sigma-Aldrich

(1S)-(−)-Camphor

95%

Synonym(s):

(−)-Camphor, (1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907612
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.24 (vs air)

Quality Level

vapor pressure

4 mmHg ( 70 °C)

Assay

95%

form

powder

autoignition temp.

870 °F

expl. lim.

3.5 %

bp

204 °C (lit.)

mp

177-179 °C (lit.)

SMILES string

[H][C@]12CC[C@](C)(C(=O)C1)C2(C)C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m0/s1

InChI key

DSSYKIVIOFKYAU-OIBJUYFYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Yamashita et al.
Pharmaceutical research, 30(1), 70-80 (2012-08-22)
Although a number of studies have reported that cocrystals can form by heating a physical mixture of two components, details surrounding heat-induced cocrystal formation remain unclear. Here, we attempted to clarify the thermal behavior of a physical mixture and cocrystal
Skrollan Stockinger et al.
Journal of chromatography. A, 1269, 346-351 (2012-08-23)
Novel 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes immobilized to poly(dimethylsiloxane) phases are presented. Immobilized 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes with a trifluoromethyl (CF(3); nickel(II)-bis[(1R,4S)-3-trifluoromethanoyl-10-propylenoxycamphor]-polysiloxane Ni(tfpc)(2)@PS) and a heptafluoropropyl-substituent (C(3)F(7); nickel(II)-bis[(1R,4S)-3-heptafluorobutanoyl-10-propylenoxycamphor]-polysiloxane Ni(hfpc)(2)@PS) were synthesized, characterized and immobilized to polysiloxane. Ni(hfpc)(2)@PS was immobilized with a selector content
Dong-Bin Dang et al.
Chemical communications (Cambridge, England), 49(22), 2243-2245 (2013-02-12)
A series of 3D homochiral manganese-lanthanide frameworks have been synthesized based on chiral camphoric acid. In the heterometallic features, D-camphoric acids are unprecedentedly embedded in three coordination modes.
[Accidental ingestion of chemical substances in the elderly].
Masashi Mise
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 25(2), 122-127 (2012-07-11)
Camphor poisoning.
K S Sahana et al.
Indian pediatrics, 49(10), 841-842 (2012-11-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service