Skip to Content
Merck
All Photos(3)

Documents

C22407

Sigma-Aldrich

3-Chloroaniline

99%

Synonym(s):

1-Amino-3-chlorobenzene, 3-Amino-1-chlorobenzene, 3-Chloro-1-aminobenzene, 3-Chlorobenzenamine, 3-Chlorophenylamine, 5-Chloroaniline, m-Aminochlorobenzene, m-Chloroaniline, m-Chlorophenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
605969
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 63.5 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.594 (lit.)

bp

95-96 °C/11 mmHg (lit.)

mp

−11-−9 °C (lit.)

density

1.206 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc(Cl)c1

InChI

1S/C6H6ClN/c7-5-2-1-3-6(8)4-5/h1-4H,8H2

InChI key

PNPCRKVUWYDDST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Elevated level of the second messenger c-di-GMP in Comamonas testosteroni enhances biofilm formation and biofilm-based biodegradation of 3-chloroaniline.: This research reveals that increased levels of c-di-GMP in Comamonas testosteroni enhance both biofilm formation and the biodegradation efficiency of 3-chloroaniline, suggesting a strategy to improve bioremediation processes (Wu et al., 2015).
  • Degradation of chloroanilines by toluene dioxygenase from Pseudomonas putida T57.: The study investigates the role of toluene dioxygenase from Pseudomonas putida T57 in degrading chloroanilines, providing insights into the enzymatic mechanisms and potential applications in treating chloroaniline pollutants (Nitisakulkan et al., 2014).
  • Bioaugmentation as a tool to protect the structure and function of an activated-sludge microbial community against a 3-chloroaniline shock load.: The study evaluates the effectiveness of bioaugmentation in maintaining the stability and functionality of microbial communities in activated sludge systems exposed to 3-chloroaniline, supporting its application in wastewater treatment (Boon et al., 2003).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1B - STOT RE 2 Oral

Target Organs

Endocrine system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Boon et al.
Applied and environmental microbiology, 67(3), 1107-1115 (2001-03-07)
We examined the diversity of the plasmids and of the gene tdnQ, involved in the oxidative deamination of aniline, in five bacterial strains that are able to metabolize both aniline and 3-chloroaniline (3-CA). Three strains have been described and identified
W Swenson et al.
Environmental microbiology, 2(5), 564-571 (2001-03-10)
We present a method for selecting entire microbial ecosystems for bioremediation and other practical purposes. A population of ecosystems is established in the laboratory, each ecosystem is measured for a desired property (in our case, degradation of the environmental pollutant
J Alary et al.
Toxicology, 59(2), 211-223 (1989-12-01)
In order to investigate the various steps of chlorpropham (CIPC) metabolism which could be influenced by cadmium, isolated rat hepatocytes were incubated in the presence of CIPC (0.1 mM) and of increasing Cd concentrations (0-180 microM). The results showed that
N Boon et al.
Applied and environmental microbiology, 66(7), 2906-2913 (2000-07-06)
A strain identified as Comamonas testosteroni I2 was isolated from activated sludge and found to be able to mineralize 3-chloroaniline (3-CA). During the mineralization, a yellow intermediate accumulated temporarily, due to the distal meta-cleavage of chlorocatechol. This strain was tested
G O Rankin et al.
Toxicology, 38(3), 269-283 (1986-03-01)
Aniline (A) and its monochlorophenyl derivatives (2-CA, 3-CA and 4-CA) are widely-used chemical intermediates. In the present study, the in vivo and in vitro nephrotoxic potential of these compounds was assessed in Fischer 344 rats. In the in vivo experiments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service