Skip to Content
Merck
All Photos(1)

Key Documents

C204

Supelco

Calmagite

metal indicator

Synonym(s):

1-(1-Hydroxy-4-methyl-2-phenylazo)-2-naphthol-4-sulfonic acid, 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)naphthalene-1-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H5[N=NC6H3(OH)CH3]SO3H
CAS Number:
Molecular Weight:
358.37
Beilstein:
708596
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Calmagite, indicator grade

grade

indicator grade

form

powder and chunks

technique(s)

titration: suitable

mp

330 °C (lit.)

λmax

602 nm

SMILES string

Cc1ccc(O)c(c1)N=Nc2c(O)cc(c3ccccc23)S(O)(=O)=O

InChI

1S/C17H14N2O5S/c1-10-6-7-14(20)13(8-10)18-19-17-12-5-3-2-4-11(12)16(9-15(17)21)25(22,23)24/h2-9,20-21H,1H3,(H,22,23,24)

InChI key

VBRNLOQCBCPPHL-UHFFFAOYSA-N

Gene Information

human ... MPL(4352)

Looking for similar products? Visit Product Comparison Guide

Application

Used as an indicator in the titration of calcium or magnesium with EDTA.
Used for the determination of magnesium in biological materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirish Agarwal et al.
Chemosphere, 73(5), 837-843 (2008-08-05)
Electrochemical degradation (ECD) is a promising technology for in situ remediation of diversely contaminated environmental matrices by application of a low level electric potential gradient. This investigation, prompted by successful bench-scale ECD of trichloroethylene, involved development, parametric characterization and evaluation
Bahram Hemmateenejad et al.
Analytica chimica acta, 607(2), 142-152 (2008-01-15)
Annihilation of the contribution of one chemical component from the original data matrix is a general method in rank annihilation factor analysis (RAFA). However, RAFA is not applicable for studying the protonation equilibria of multiprotic acids. In this work, a
Calmagite method for determination of serum magnesium modified.
S A Cohen et al.
Clinical chemistry, 26(6), 783-783 (1980-05-01)
Rapid measurement of serum magnesium with a kit.
P West
Clinical chemistry, 30(8), 1426-1427 (1984-08-01)
Laboratory Medicine, 12, 139-139 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service