Key Documents
BR136100
BRAND® stirring spatula
high-density polyethylene
Synonym(s):
Stirring rods
About This Item
UNSPSC Code:
41122403
NACRES:
NB.72
Pricing and availability is not currently available.
material
high-density polyethylene
description
with steel core
packaging
pack of 1 ea
manufacturer/tradename
BRAND 136100
L × D
260 mm × 7 mm
General description
The BRAND® stirring spatula features a PE-HD coated steel core, providing durability and strength for various laboratory applications. With two differently shaped ends, each approximately 16 mm wide, this spatula is designed for effective mixing, stirring, and scraping of both solid and liquid substances. The PE-HD coating ensures resistance to chemicals and prevents contamination during use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)