BAF170720010
Cleanware microscope optics, additional optical cleaning pads
pack of 10 ea
All Photos(1)
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
Recommended Products
packaging
pack of 10 ea
manufacturer/tradename
Bel-Art F17072-0010
General description
Contains everything you need to keep optics and microscope parts sanitary, clean and clear. Ten pre-saturated optical cleaning pads, two individually wrapped packs of anti-static plastic surface wipes and Two, 15.2cm (6") swabs pre-moistened with alcohol, contains complete instructions for use.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)