Skip to Content
Merck
All Photos(3)

Documents

B9807

Sigma-Aldrich

1,1,2,2-Tetraphenyl-1,2-ethanediol

99%

Synonym(s):

TPED, Benzopinacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C(OH)C(C6H5)2OH
CAS Number:
Molecular Weight:
366.45
Beilstein:
2061947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

171-173 °C (lit.)

SMILES string

OC(c1ccccc1)(c2ccccc2)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C26H22O2/c27-25(21-13-5-1-6-14-21,22-15-7-2-8-16-22)26(28,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20,27-28H

InChI key

MFEWNFVBWPABCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 1,1,2,2-Tetraphenyl-1,2-ethanediol (TPED) is used as an active component of the initiation system in free radical polymerization.
  • It is used as a precursor to prepare 1-hydroxy-2-(p-tolylaminocarbonyloxy)-1,1,2,2-tetraphenylethane (HTTPE), a thermal iniferter for radical polymerization.
  • TPED can also be used as a protecting group for the boronic acids by converting them to stable boronic esters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 1, 2, 2-Tetraphenyl-1, 2-ethanediol: A new protecting group for the synthesis of cyclopropylboronic esters.
Pietruszka J and Witt A
Synlett, 2003(01), 0091-0094 (2003)
Synthesis of well?defined polystyrene by radical polymerization using 1, 1, 2, 2?tetraphenyl?1, 2?ethanediol/feCl3/PPh3 initiation system.
Chen X P and Qiu K Y
Journal of Applied Polymer Science, 77(7), 1607-1613 (2000)
A novel thermal iniferter for radical polymerization.
Chen X P, et al.
European Polymer Journal, 36(8), 1547-1554 (2000)
Synthesis of well-defined poly (methyl methacrylate) by radical polymerization with a new initiation system TPED/FeCl3/PPh3.
Chen X P and Qiu K Y
Macromolecules, 32(26), 8711-8715 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service