Skip to Content
Merck
All Photos(1)

Key Documents

B83207

Sigma-Aldrich

5-Bromovaleronitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)4CN
CAS Number:
Molecular Weight:
162.03
Beilstein:
1742080
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.478 (lit.)

density

1.388 g/mL at 25 °C (lit.)

SMILES string

BrCCCCC#N

InChI

1S/C5H8BrN/c6-4-2-1-3-5-7/h1-4H2

InChI key

NWWWGAKVHCSAEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromovaleronitrile can be used as a reactant to prepare:
  • A key intermediate benzyl(methyl)amino)pentanenitrile, which is used in the synthesis of N-methylcadaverine.
  • 1-cyanobutyl-3-alkylbenzimidazolium bromide salt by reacting with various N-alkylbenzimidazoles.
  • Tridecanenitrile by Ni-catalyzed cross-coupling reaction with dioctylzinc in the presence of tetraene and MgBr2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle Muñoz-Osses et al.
Dalton transactions (Cambridge, England : 2003), 47(4), 1233-1242 (2018-01-05)
Substituted amino-piperazine derivatives were synthesized and used as precursors for the preparation of a series of new organometallic Re(i) imine complexes with the general formula [(η
Nitrile functionalized silver (I) N-heterocyclic carbene complexes: DFT calculations and antitumor studies
Hussaini SY, et al.
Transition Metal Chemistry, 43(4), 301-312 (2018)
Kayla N Anderson et al.
Molecules (Basel, Switzerland), 23(5) (2018-05-23)
Alkaloids compose a large class of natural products, and mono-methylated polyamines are a common intermediate in their biosynthesis. In order to evaluate the role of selectively methylated natural products, synthetic strategies are needed to prepare them. Here, N-methylcadaverine is prepared
Nickel-catalyzed cross-coupling reaction of alkyl halides with organozinc and Grignard reagents with 1,3,8,10-tetraenes as additives.
Jun Terao et al.
Angewandte Chemie (International ed. in English), 43(45), 6180-6182 (2004-11-19)
Franziska Kupke et al.
Scientific reports, 6, 37631-37631 (2016-11-25)
Isothiocyanates are the most intensively studied breakdown products of glucosinolates from Brassica plants and well recognized for their pleiotropic effects against cancer but also for their genotoxic potential. However, knowledge about the bioactivity of glucosinolate-borne nitriles in foods is very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service