Skip to Content
Merck
All Photos(1)

Key Documents

B62404

Sigma-Aldrich

1-Bromo-3-chloropropane

99%

Synonym(s):

I-BCP, Trimethylene bromochloride, Trimethylene chlorobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3Br
CAS Number:
Molecular Weight:
157.44
Beilstein:
605278
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.486 (lit.)

bp

144-145 °C (lit.)

density

1.592 g/mL at 25 °C (lit.)

SMILES string

ClCCCBr

InChI

1S/C3H6BrCl/c4-2-1-3-5/h1-3H2

InChI key

MFESCIUQSIBMSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Bromo-3-chloropropane is generally used as C3 linker/spacer in a wide range of molecules including medicinally important compounds. Some of the examples are:
  • Synthesis of polymer nanoparticles (NPs) to deliver doxorubicin, camptothecin, and cisplatin.
  • Synthesis of antiprion active acridine derivatives for prion infection.
  • Total synthesis of aspidospermine, aspidospermidine, and quebrachamine and (±)-crispine A.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient synthesis of tricyclic amines by a cyclization/cycloaddition cascade: total syntheses of aspidospermine, aspidospermidine, and quebrachamine.
Coldham, Iain et al.
Angewandte Chemie (International Edition in English), 46(32), 6159-6162 (2007)
A chimeric ligand approach leading to potent antiprion active acridine derivatives: design, synthesis, and biological investigations.
Dollinger, Silke et al.
Journal of Medicinal Chemistry, 49(22), 6591-6595 (2006)
Design, synthesis, and evaluation of novel fluoroquinolone- aminoglycoside hybrid antibiotics.
Pokrovskaya, Varvara et al.
Journal of Medicinal Chemistry, 52(8), 2243-2254 (2009)
Discovery of novel and potent thiazoloquinazolines as selective Aurora A and B kinase inhibitors.
Jung, Frederic H et al.
Journal of Medicinal Chemistry, 49(3), 955-970 (2006)
Cascade condensation, cyclization, intermolecular dipolar cycloaddition by multi-component coupling and application to a synthesis of (?)-crispine A.
Coldham, Iain et al.
Organic & Biomolecular Chemistry, 7(8), 1674-1679 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service