Skip to Content
Merck
All Photos(1)

Key Documents

B59608

Sigma-Aldrich

α-Bromo-γ-butyrolactone

97%

Synonym(s):

2-Bromo-4-hydroxybutyric acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5BrO2
CAS Number:
Molecular Weight:
164.99
Beilstein:
107800
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

138 °C/6 mmHg (lit.)

density

1.786 g/mL at 25 °C (lit.)

SMILES string

BrC1CCOC1=O

InChI

1S/C4H5BrO2/c5-3-1-2-7-4(3)6/h3H,1-2H2

InChI key

LFJJGHGXHXXDFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service