Key Documents
ALDKIT001
Sigma-Aldrich
Aldrich® Micro Photochemical Reactor, blue LED lights
AC/DC input 100 V / 240 V AC, universal plug set
Synonym(s):
Micro photochemical reactor
About This Item
UNSPSC Code:
41106514
NACRES:
NC.25
Pricing and availability is not currently available.
AC/DC input
(US/UK/Australia /Scandanavia AC interchangable input plugs)
feature
capacity 16  vial(s)
parameter
435-445 nm spectral range
General description
This newly designed apparatus enables researchers to process up to 16 samples at once. Safe LED light provides the energy needed for the photoreactions.
Features and Benefits
  • Aluminum sample base with riser feet holds up to 16 samples and sits atop a magnetic stirrer (5 X 7 plate size or larger recommended but not included)
  • LED light ring is IP68 double density 12vdc water proof blue, 435-445 nm
  • Uses 4 mL sample vials with either PTFE lined solid top caps or holed caps with PTFE face rubber septa (purchase separately)
  • Power supply is wall plug power supply 500 mA 5-6 watts
  • Use with stir bars Z118850 (10 x 3 mm)
  • Use with 4 mL solid top capped vials 27138 or septum capped vials 27209-U
  • Optional purple LED light ring available ALDRP2, 400-410nm. (Light is 1/2 the brightness of blue - MORE UV REACTIVE}
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research.
Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research.
Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research.
Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research.
See All
Related Content
Yoon lab exploits visible light photochemistry for unique chemical transformations.
Yoon lab exploits visible light photochemistry for unique chemical transformations.
Questions
1–3 of 3 Questions  
  1. Guten Tag, wir haben uns gefragt, wie hoch die Optical Output Power der LED (in mW) ist.
    1 answer
    1. The LED power is 0.3W or 300 mW per sample vial.
      Helpful?
  2. How can I determine the shelf life / expiration / retest date of this product?
    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
      Helpful?
  3. How is shipping temperature determined? And how is it related to the product storage temperature?
    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
      Helpful?