Skip to Content
Merck
All Photos(3)

Key Documents

A79922

Sigma-Aldrich

9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate

≥99%

Synonym(s):

THA hydrochloride hydrate, Tacrine hydrochloride, Tetrahydroaminacrine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2 · HCl · xH2O
CAS Number:
Molecular Weight:
234.72 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99%

form

solid

mp

284-286 °C (lit.)

SMILES string

O.Cl.Nc1c2CCCCc2nc3ccccc13

InChI

1S/C13H14N2.ClH.H2O/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13;;/h1,3,5,7H,2,4,6,8H2,(H2,14,15);1H;1H2

InChI key

PXGRMZYJAOQPNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate is a well-known cholinesterase inhibitor commonly used in pharmacological studies.[1][2][3][4][5]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetylcholinesterase capillary enzyme reactor for screening and characterization of selective inhibitors.
da Silva JI, et al.
Journal of Pharmaceutical and Biomedical Analysis, 73, 44-52 (2013)
The ameliorating effects of stigmasterol on scopolamine-induced memory impairments in mice.
Park SJ, et al.
European Journal of Pharmacology, 676(1-3), 64-70 (2012)
Comparison of inhibitory activities of donepezil and other cholinesterase inhibitors on acetylcholinesterase and butyrylcholinesterase in vitro.
Ogura H, et al.
Methods and Findings in Experimental and Clinical Pharmacology, 22(8), 609-614 (2000)
A review on cholinesterase inhibitors for Alzheimer?s disease.
Anand P and Singh B
Archives of Pharmacal Research, 36(4), 375-399 (2013)
New Tacrines as Anti-Alzheimer's Disease Agents. The (Benzo) Chromeno-PyranoTacrines.
Oset-Gasque MJ and Marco-Contelles J
Current Topics in Medicinal Chemistry, 17(31), 3349-3360 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service