Skip to Content
Merck
All Photos(1)

Key Documents

A45807

Sigma-Aldrich

2-Amino-5-chlorobenzoxazole

97%

Synonym(s):

Zoxazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClN2O
CAS Number:
Molecular Weight:
168.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

181-184 °C (lit.)

SMILES string

Nc1nc2cc(Cl)ccc2o1

InChI

1S/C7H5ClN2O/c8-4-1-2-6-5(3-4)10-7(9)11-6/h1-3H,(H2,9,10)

InChI key

YGCODSQDUUUKIV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huidi Jiang et al.
Die Pharmazie, 58(9), 654-656 (2003-10-09)
The present paper aims to investigate whether p-amino-2',4'-dichlorodiphenyl ether and p-amino-4'-methyldiphenyl ether are inhibitors as well as inducers of P450. Mice were given daily intraperitoneal (ip) injections of p-amino-2',4'-dichlorodiphenyl ether (0.25 mmol/kg) or p-amino-4'-methyldiphenyl ether (0.25 mmol/kg) for 4 days
Romi Ghose et al.
Life sciences, 89(1-2), 57-64 (2011-05-31)
Our aim is to investigate the molecular mechanism of regulation of gene expression of drug metabolizing enzymes (DMEs) and transporters in diet-induced obesity. Adult male CD1 mice were fed diets containing 60% kcal fat (HFD) or 10% kcal fat (LFD)
Yuzhi Jia et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(35), 12531-12536 (2005-08-20)
Peroxisome proliferator-activated receptor-binding protein (PBP), also known as thyroid hormone receptor-associated protein 220/vitamin D receptor-interacting protein 205/mediator 1, an anchor for multisubunit mediator transcription complex, functions as a transcription coactivator for nuclear receptors. Disruption of the PBP gene results in
W Tracewell et al.
Toxicology and applied pharmacology, 135(2), 179-184 (1995-12-01)
CYP1A1 gene expression is regulated by known cis- and transacting elements controlling inhibition and induction of CYP1A1 transcription. The influence of a double-stranded phosphorothioate oligonucleotide (dsODN) with sequence identical to the CYP1A1 negative regulatory element (NRE) was examined in Sprague-Dawley
Juan P Hernandez et al.
Toxicology and applied pharmacology, 216(2), 186-196 (2006-07-11)
Nonylphenol (NP) is a breakdown product of nonylphenol ethoxylates, which are used in a variety of industrial, agricultural, household cleaning, and beauty products. NP is one of the most commonly found toxicants in the United States and Europe and is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service