Skip to Content
Merck
All Photos(2)

Documents

A16602

Sigma-Aldrich

DL-N-Acetylhomocysteine thiolactone

98%

Synonym(s):

2-Acetamido-4-mercaptobutyric acid γ-thiolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO2S
CAS Number:
Molecular Weight:
159.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

109-111 °C (lit.)

SMILES string

CC(=O)NC1CCSC1=O

InChI

1S/C6H9NO2S/c1-4(8)7-5-2-3-10-6(5)9/h5H,2-3H2,1H3,(H,7,8)

InChI key

NRFJZTXWLKPZAV-UHFFFAOYSA-N

Application

<ul>
<li><strong>Ultrasonic synthesis of stable oil filled microcapsules:</strong> This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Powers et al.
Endocrinology, 137(7), 3101-3110 (1996-07-01)
The activity of nitric oxide (NO) has been implicated as an important mediator in ovarian function, including the regulation of the blood-follicle barrier and ovulation. Both clinical and experimental diabetes have been found to impair endothelial-dependent vascular activity through the
P P Sood et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 14(2), 101-105 (1995-01-01)
Male albino mice were intoxicated with a daily dose of 1 mg/kg of methylmercury chloride (MMC) for 7 days, and were treated thereafter with glutathione, N-acetyl-DL-homocysteine thiolactone, vitamin B complex, and vitamin E, either alone or in combinations, for the
I Flechner et al.
Diabetes research (Edinburgh, Scotland), 13(2), 67-73 (1990-02-01)
The possible value of radical scavengers in the prevention of beta cell destruction was studied in two animal models of type I diabetes. Eight compounds were tested alone or in combinations. In the low-dose streptozotocin model treatment started 24 hr
Fixed drug eruption due to citiolone.
P G Delgado et al.
Contact dermatitis, 33(5), 352-352 (1995-11-01)
W Sumoski et al.
Diabetologia, 32(11), 792-796 (1989-11-01)
A possible role for oxygen free radicals in mediating the cytotoxic effects of cytokines in islets was sought by the use of agents that scavenge free radicals. Rat islet cell monolayer cultures were incubated for 6 days with t-butylhydroperoxide, alloxan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service