Skip to Content
Merck
All Photos(2)

Key Documents

938742

Sigma-Aldrich

RuPhos Pd G6 Br

Synonym(s):

(RuPhos)Pd(p-CF3C6H4)(Br), 2-Dicyclohexylphosphino-2′,6′-diisopropoxybiphenyl Pd G6 Br

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H47BrF3O2PPd
CAS Number:
Molecular Weight:
798.06
UNSPSC Code:
12352100
NACRES:
NA.21

form

powder

Quality Level

reaction suitability

reaction type: Cross Couplings
reagent type: catalyst

color

very faintly yellow

functional group

phosphine

SMILES string

Br[Pd](C1=CC=C(C=C1)C(F)(F)F)P(C2=C(C3=C(C=CC=C3OC(C)C)OC(C)C)C=CC=C2)(C4CCCCC4)C5CCCCC5

InChI

InChI=1S/C30H43O2P.C7H4F3.BrH.Pd/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25;8-7(9,10)6-4-2-1-3-5-6;;/h11-13,18-25H,5-10,14-17H2,1-4H3;2-5H;1H;/q;;;+1/p-1

InChI key

IYBINOQILDORLG-UHFFFAOYSA-M

General description

RuPhos Pd G6 is a G6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.

Features and Benefits

For feature and benefits, please visit the technical article on Buchwald G6 Precatalysts.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service