Skip to Content
Merck
All Photos(1)

Key Documents

935530

Sigma-Aldrich

Silver(I) chloride

-325 mesh, 99.99% trace metals basis (excluding Na)

Synonym(s):

Silver chloride, Chlorosilver, Silver monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
MDL number:
UNSPSC Code:
12352302

vapor pressure

1 mmHg ( 912 °C)

Quality Level

Assay

73.4-77.1% (with AgNO3, titration)
99.99% trace metals basis (excluding Na)

form

powder

impurities

≤150 ppm (trace metals analysis)

color

white to off-white

particle size

-325 mesh

mp

455 °C (lit.)
455 °C

solubility

water: soluble

SMILES string

Cl[Ag]

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver(I) chloride is a off-white solid which is slightly soluble in water. Silver(I) chloride turns black in the presence of heat, indicating the formation of silver.
It can be produced easily through the combination of silver nitrate, sodium chloride, or cobalt(II) chloride.

Application

Silver chloride (AgCl) is widely employed as a catalyst, antibacterial agent, photographic material, and ionic semiconductor material.
Silver chloride (AgCl) can be used for a variety of applications such as: the coating on the silver wire that can be used as a reference electrode in electrochemistry
It can be deposited on conducting material for stable photocatalysis under visible light .
In addition, Silver(I)chloride is used to synthesis polymeric silver(I) complexes with 1D and 3D network .
Moreover, Silver(I)chloride is also used as a catalyst in the preparation of 1,2,3-triazoles from alkynes and azides.

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitivities of key parameters in the preparation of silver/silver chloride electrodes used in Harned cell measurements of pH
Paul J, et al.
Sensors (Basel, Switzerland), 11(8), 8072-8084 (2011)
Controlled Assembly Based on Multibridging Thiolate Ligands: New Polymeric Silver(I) Complexes with One-Dimensional Chain and Three-Dimensional Network Structures.
Hong M, et al.
Inorganic Chemistry, 38(3), 600-602 (1999)
Ag@ AgCl: a highly efficient and stable photocatalyst active under visible light.
Wang P, et al.
Angewandte Chemie (International Edition in English), 47(41), 7931-7933 (2008)
Fabrication and characterization of visible-light-driven plasmonic photocatalyst Ag/AgCl/TiO2 nanotube arrays.
Yu J, et al.
The Journal of Physical Chemistry C, 113(37), 16394-16401 (2009)
The silver chloride photoanode in photoelectrochemical water splitting
Schurch D, et al.
The Journal of Physical Chemistry B, 106(49), 12764-12775 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service