926558
Upconversion Nanoparticles
NaYF4-Yb,Tm@NaYF4, PEG-NH2 modified core-shell ucnps,980 excitation, blue light
Synonym(s):
UCNPs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
concentration
2 mg/mL in water
matrix active group
PEG-NH2 surface treatment
particle size
30 nm±5 nm
fluorescence
λex 980 nm (blue)
λem 360 nm
λem 450 nm
λem 470 nm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Fluorescent microscopy
- Deep-tissue bioimaging
- Nanomedicine
- Optogenetics
- Security labelling
- Volumetric display
Upconversion nanoparticles (ucnps), are the result of a unique optical process in which near-infrared (NIR) light excitation is converted into visable and/or UV emission. Upconversion (UC) utilizes sequential absorption of multiple photons utilizing both lifetime and real ladder-like energy levels of ions in a host lattice to produce a higher energy anti-Stokes luminescence.
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
Legal Information
Product of RuixiBiotechCo. Ltd
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amplified stimulated emission in upconversion nanoparticles for super-resolution nanoscopy
Nature, 543, 229?233 -229?233 (2017)
Engineering bright sub-10-nm upconverting nanocrystals for single-molecule imaging
Nature Nanotechnology, 9, 300?305-300?305 (2014)
Advances in highly doped upconversion nanoparticles
Nature Communications, 9, 2415-2415 (2018)
Engineered upconversion nanoparticles for resolving protein interactions inside living cells
Angewandte Chemie (International ed. in English), 55, 11668?11672-11668?11672 (2016)
Upconversion nanoparticles: design, nanochemistry, and applications in theranostics
Chemical Reviews, 114, 5161-5214 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service