Skip to Content
Merck
All Photos(3)

Key Documents

926272

Sigma-Aldrich

Stir Bar Actuated Mixer Chip

Fluidic 286, COP

Synonym(s):

Microfluidic chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

description

Microfludic chip x1

Looking for similar products? Visit Product Comparison Guide

Application

The Stir Bar Actuated Mixer Fluidic 286 COP microfluidic chip applies active mixing principles by integrating stir bars to generate mixtures with a wider range of mixing ratios that are not feasible with passive mixing devices.

Features and Benefits

  • Six micromixing units varying from 20 to 150 μl chamber volumes
  • Each unit features two inlets and one outlet
  • Can generate mixtures with a wide range of mixing ratios (ie 1:10)
  • Can be powered on any standard magnetic stir plate

Components

Chip Properties:
  • Mini Luer Interface
  • Material: Cyclic olefin polymer (COP)
  • Chamber Volumes : 20 μl, 40 μl, 60 μl, 80 μl, 100 μl, 150 μl

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired

Articles

Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.

Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service