Skip to Content
Merck
All Photos(2)

Key Documents

909602

Sigma-Aldrich

DSSO crosslinker

≥95%

Synonym(s):

Bis(2,5-dioxopyrrolidin-1-yl) 3,3′-sulfinyldipropionate, Bis-(propionic acid NHS ester)-sulfoxide, Mass spectrometry-cleavable crosslinker for studying protein-protein interations

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O9S
CAS Number:
Molecular Weight:
388.35
MDL number:
UNSPSC Code:
12161502

Assay

≥95%

form

powder

availability

available only in USA

storage temp.

2-8°C

SMILES string

[S](=O)(CCC(=O)ON2C(=O)CCC2=O)CCC(=O)ON1C(=O)CCC1=O

InChI key

XJSVVHDQSGMHAJ-UHFFFAOYSA-N

Application

DSSO (disuccinimidyl sulfoxide) crosslinker is a homobifunctional, amine-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). Membrane-permeable DSSO possesses two N-hydroxysuccinimide (NHS) esters for targeting Lys, a 10.1 Å spacer arm, and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. The post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS. DSSO Crosslinker provides complementary data to thiol-reactive and acidic residue-targeting reagents and will find wide utility in the elucidation of PPIs, study of proteins that function as complexes, quantification of structural dynamics, and the quest for targeting ″undruggable″ protein targets.

Legal Information

Subject to US Patent #9,222,943 and US Patent Application #15/275,001 of the Regents of the University of California

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian E Stieger et al.
Journal of proteome research, 18(3), 1363-1370 (2019-01-30)
Cross-linking mass spectrometry is becoming increasingly popular, and current advances are widening the applicability of the technique so that it can be utilized by nonspecialist laboratories. Specifically, the use of novel mass-spectrometry-cleavable (MS-cleavable) reagents dramatically reduces the complexity of the
Sebastiaan C de Graaf et al.
Journal of proteome research, 18(2), 642-651 (2018-12-24)
Protein interactions enable much more complex behavior than the sum of the individual protein parts would suggest and represents a level of biological complexity requiring full understanding when unravelling cellular processes. Cross-linking mass spectrometry has emerged as an attractive approach
Athit Kao et al.
Molecular & cellular proteomics : MCP, 10(1), M110-M110 (2010-08-26)
Knowledge of elaborate structures of protein complexes is fundamental for understanding their functions and regulations. Although cross-linking coupled with mass spectrometry (MS) has been presented as a feasible strategy for structural elucidation of large multisubunit protein complexes, this method has
Daniela-Lee Smith et al.
Analytical chemistry, 90(15), 9101-9108 (2018-07-14)
This study investigated the enzyme-substrate interaction between Saccharomyces cerevisiae arginine methyltransferase Hmt1p and nucleolar protein Npl3p, using chemical cross linking/mass spectrometry (XL/MS). We show that XL/MS can capture transient interprotein interactions that occur during the process of methylation, involving a
Tara K Bartolec et al.
Analytical chemistry, 92(2), 1874-1882 (2019-12-19)
Saccharomyces cerevisiae has the most comprehensively characterized protein-protein interaction network, or interactome, of any eukaryote. This has predominantly been generated through multiple, systematic studies of protein-protein interactions by two-hybrid techniques and of affinity-purified protein complexes. A pressing question is to

Articles

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service