Skip to Content
Merck
All Photos(1)

Key Documents

905623

Sigma-Aldrich

Gold nanospheres

15 nm, Cy3 and amine functionalized, powder

Synonym(s):

Cy3 labeled gold nanospheres, Cy3 tagged gold nanospheres, Dye labeled gold nanoparticles, Fluorescent labeled gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

nanospheres
powder

particle size

15 nm

λmax

554 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanospheres (AuNS) (100nm, Cy3 and amine functionalized, powder) are gold nanoparticles with unique properties which include biocompatibility, easy surface modification, and excellent photothermal conversion. They can be synthesized by a variety of methods such as galvanic replacement of gold nanoparticles, templated galvanic replacement of silver for gold, and electrochemical methods.[1]

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties[2][3][4] which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy[2][3][4][5][6][7]. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores[8]. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications[2][8][9].

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, modification, and application of hollow gold nanospheres
Ren Q, et al.
Journal of Nanomaterials, 2015 (2015)
Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching.
Lu Y, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Gold nanoparticles in chemical and biological sensing.
Krishnendu Saha et al.
Chemical reviews, 112(5), 2739-2779 (2012-02-03)
Nuri Oh et al.
ACS nano, 8(6), 6232-6241 (2014-05-20)
Significant quantities of synthetic nanoparticles circulating in the body are cleared and retained for long periods of time in the resident macrophages of the mononuclear phagocytic system (MPS), increasing the likelihood of nanoparticle-mediated chronic toxicity. To date, there has been
Byoungjin Kim et al.
Nature nanotechnology, 5(6), 465-472 (2010-04-13)
Nanoparticles have great potential as controllable drug delivery vehicles because of their size and modular functionality. Timing and location are important parameters when optimizing nanoparticles for delivery of chemotherapeutics. Here, we show that gold nanoparticles carrying either fluorescein or doxorubicin

Articles

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service