Skip to Content
Merck
All Photos(1)

Documents

905615

Sigma-Aldrich

Gold nanospheres

100 nm, FITC and amine functionalized, powder

Synonym(s):

Dye labeled gold nanoparticles, FITC labeled gold nanospheres, FITC tagged gold nanospheres, Fluorescein isothiocyanate labeled gold nanospheres, Fluorescent labeled gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanospheres
powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanospheres (AuNS) (100nm, FITC and amine functionalized, powder) are gold nanoparticles with unique properties which include biocompatibility, easy surface modification, and excellent photothermal conversion. They can be synthesized by a variety of methods such as galvanic replacement of gold nanoparticles, templated galvanic replacement of silver for gold, and electrochemical methods.

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy.The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores.This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, modification, and application of hollow gold nanospheres
Ren Q, et al.
Journal of Nanomaterials, 2015 (2015)
Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching
Y. Lu et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching.
Lu Y, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Byoungjin Kim et al.
Nature nanotechnology, 5(6), 465-472 (2010-04-13)
Nanoparticles have great potential as controllable drug delivery vehicles because of their size and modular functionality. Timing and location are important parameters when optimizing nanoparticles for delivery of chemotherapeutics. Here, we show that gold nanoparticles carrying either fluorescein or doxorubicin
Gold nanoparticles: assembly, supramolecular chemistry, quantum-size-related properties, and applications toward biology, catalysis, and nanotechnology.
Marie-Christine Daniel et al.
Chemical reviews, 104(1), 293-346 (2004-01-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service