Skip to Content
Merck
All Photos(2)

Key Documents

901235

Sigma-Aldrich

TBS-DHG Catalyst

greener alternative

≥95%

Synonym(s):

(2R,3S,4R)-2-(((tert-Butyldimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-3,4-diol, 6-Tertbutyldimethylsilyl-1,2-dihydroglucal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26O4Si
CAS Number:
Molecular Weight:
262.42
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

SMILES string

[H]C1([H])C([H])([H])O[C@@](C([H])([H])O[Si](C([H])([H])[H])(C([H])([H])[H])C(C([H])([H])[H])(C([H])([H])[H])C([H])([H])[H])([H])[C@](O[H])([H])[C@]1([H])O[H]

InChI

1S/C12H26O4Si/c1-12(2,3)17(4,5)16-8-10-11(14)9(13)6-7-15-10/h9-11,13-14H,6-8H2,1-5H3/t9-,10-,11+/m1/s1

InChI key

KKFGQPZYDWCKRC-MXWKQRLJSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

6-Tertbutyldimethylsilyl-1,2-dihydroglucal (TBS-DHG) was reported by the Morken Lab to be an effective carbohydrate-derived catalyst for enantioselective diboration of alkenes. Related capabilities were observed with the dihydrorhamnal (DHR) catalyst (901237). TBS-DHG has also been used in ruthenium(0)catalyzed transfer hydrogenation cycloadditions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Sato et al.
Journal of the American Chemical Society, 138(50), 16244-16247 (2016-12-10)
The first use of vicinal diols, ketols, or diones as 2
Zachary A Kasun et al.
Chemical communications (Cambridge, England), 50(56), 7545-7547 (2014-06-04)
A new method for the ring expansion of cyclic diols is described. Using improved conditions for the ruthenium(0) catalyzed cycloaddition of cyclic 1,2-diols with 1,3-dienes, fused [n.4.0] bicycles (n = 3-6) are formed, which upon exposure to iodosobenzene diacetate engage
Lichao Fang et al.
Journal of the American Chemical Society, 138(8), 2508-2511 (2016-02-09)
Catalytic enantioselective diboration of alkenes is accomplished with readily available carbohydrate-derived catalysts. Mechanistic experiments suggest the intermediacy of 1,2-bonded diboronates.
Lu Yan et al.
Journal of the American Chemical Society, 140(10), 3663-3673 (2018-02-15)
A mechanistic investigation of the carbohydrate/DBU cocatalyzed enantioselective diboration of alkenes is presented. These studies provide an understanding of the origin of stereoselectivity and also reveal a strategy for enhancing reactivity and broadening the substrate scope.

Related Content

Chiral organoboronic esters are well known as versatile intermediates for chemical synthesis. Not only are these compounds stable under a variety of reaction conditions, they are generally non-toxic and can be transformed with minimal generation of hazardous waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service