Skip to Content
Merck
All Photos(1)

Key Documents

900683

Sigma-Aldrich

2-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)ethan-1-amine

95%

Synonym(s):

Amino diazirine alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12352116
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

storage temp.

−20°C

SMILES string

NCCC1(N=N1)CCC#C

InChI key

FMCIBERQJQEYIZ-UHFFFAOYSA-N

Application

2-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)ethan-1-amine is used for chemical probe synthesis, this trifunctional building block contains a light-activated diazirine, alkyne tag, and amine synthetic handle. When appended to a ligand or pharmacophore through its amine linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service