Skip to Content
Merck
All Photos(2)

Key Documents

900442

Sigma-Aldrich

Graphene/PEDOT:PSS hybrid ink

dispersion in DMF, avg. no. of layers, 1 ‑ 3

Synonym(s):

Conductive ink, Electrochemically exfoliated graphene ink, G/PEDOT:PSS, Graphene ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

dispersion (in DMF)

feature

avg. no. of layers 1 ‑ 3

concentration

0.2 mg/mL (PEDOT:PSS)
1 mg/mL (electrochemically exfoliated graphene)

sheet resistance

500 Ω/sq, 20 nm film: 80% transmittance

General description

  • Graphene preparation method: Electrochemical exfoliation.
  • Graphene thickness by AFM: 80%, 1-3 layers.
  • Sheet size by AFM: 10 μm.
  • Oxygen content: 7.5% (by XPS) (C/O-ratio: 12.3).
  • Raman I_D/I_G ratio: 0.4.
  • Sheet resistance: 4.8 kΩ/sq.
  • Typical properties of films produced thereof (after spray coating):
a) 11 nm film: 90% transmittance, 1200 Ω/sq (as made).
b) 20 nm film: 80% transmittance, 500 Ω/sq (as made).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F

Flash Point(C)

58 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoyang Liu et al.
Advanced materials (Deerfield Beach, Fla.), 27(4), 669-675 (2014-12-03)
A novel solution fabrication of large-area, highly conductive graphene films by spray-coating of a hybrid ink of exfoliated graphene (EG)/poly(3,4-ethylenedioxythiophene):poly(styrenesulfonate) (PEDOT:PSS) (PH1000) is demonstrated. The fabricated graphene films exhibit excellent mechanical properties, thus enabling their application as bottom electrodes in
Transparent graphene/PEDOT-PSS composite films as counter electrodes of dye-sensitized solar cells.
Hong, et al.
Electrochemical Communications, 10(10), 1555-1558 (2008)
Peng You et al.
Advanced materials (Deerfield Beach, Fla.), 27(24), 3632-3638 (2015-05-15)
Semitransparent perovskite solar cells are prepared by laminating graphene transparent electrodes on the top for the first time. The device performance is optimized by improving the conductivity of the graphene electrodes and the contact between the graphene and the perovskite
Zhong-Shuai Wu et al.
Advanced materials (Deerfield Beach, Fla.), 27(24), 3669-3675 (2015-05-15)
Ultrathin printable graphene supercapacitors are demonstrated, based on solution-processed electrochemically exfoliated graphene hybrid films on an ultrathin poly(ethylene terephthalate) substrate, exhibiting an unprecedented volumetric capacitance of 348 F cm(-3) , an ultrahigh scan rate of 2000 V s(-1) , and

Articles

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service