Skip to Content
Merck
All Photos(1)

Documents

859885

Sigma-Aldrich

(Z)-9-Tricosene

97%

Synonym(s):

cis-9-Tricosene, Muscalure

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH=CH(CH2)7CH3
CAS Number:
Molecular Weight:
322.61
Beilstein:
1841622
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

300 °C (lit.)

density

0.806 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCCCC

InChI

1S/C23H46/c1-3-5-7-9-11-13-15-17-19-21-23-22-20-18-16-14-12-10-8-6-4-2/h17,19H,3-16,18,20-23H2,1-2H3/b19-17-

InChI key

IGOWHGRNPLFNDJ-ZPHPHTNESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Tillman-Wall et al.
Archives of biochemistry and biophysics, 299(1), 92-99 (1992-11-15)
The regulation of production of the sex pheromone (Z)-9-tricosene (Z9-23:Hy) in the housefly, Musca domestica, was studied by examining the chain length specificity of the fatty acyl-CoA elongation reactions and the reductive conversion of fatty acyl-CoAs to alkenes in 1-
Sarah M Butler et al.
Journal of economic entomology, 100(4), 1489-1495 (2007-09-14)
Sticky card captures of house flies, Musca domestica L. (Diptera: Muscidae), were used to compare efficacy of screen-covered baits containing sugar, sugar and 0.1% (Z)-9-tricosene, sugar and 1.0% (Z)-9-tricosene, Golden Malrin [1.1% methomyl and 0.049% (Z)-9-tricosene], and Quick-Bayt [0.5% imidacloprid
J W Chapman et al.
Journal of economic entomology, 91(4), 915-922 (1998-09-02)
A field trial comparing the effectiveness of toxic targets impregnated with different formulations of the Musca domestica L. female sex pheromone (Z)-9-tricosene was conducted in a caged-layer, deep-pit poultry unit in southern England. Targets baited with 5 g of technical
Mihaela Kavran et al.
Annals of agricultural and environmental medicine : AAEM, 22(2), 243-246 (2015-06-23)
Flies - by feeding on decaying matter, human waste and food - have been implicated in the spread of numerous animal and human diseases. Excessive fly populations are generally associated with livestock units and domestic waste due to decaying organic
J W Chapman et al.
Medical and veterinary entomology, 13(2), 132-138 (1999-09-14)
Field trials investigating the effect of visual cues on catches of Musca domestica (Diptera: Muscidae) at toxic targets impregnated with the female sex pheromone (Z)-9-tricosene, were conducted in a caged-layer deep-pit poultry unit in southern England. Targets treated with azamethiphos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service