Skip to Content
Merck
All Photos(3)

Key Documents

858781

Sigma-Aldrich

3,4-Dihydroxybenzylamine hydrobromide

98%

Synonym(s):

4-(Aminomethyl)catechol hydrobromide, DHBA hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2NH2 · HBr
CAS Number:
Molecular Weight:
220.06
Beilstein:
4002646
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

crystals

mp

184-186 °C (lit.)

functional group

amine

SMILES string

Br.NCc1ccc(O)c(O)c1

InChI

1S/C7H9NO2.BrH/c8-4-5-1-2-6(9)7(10)3-5;/h1-3,9-10H,4,8H2;1H

InChI key

BVFZTXFCZAXSHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Oxidative Polymerization of 3,4-Dihydroxybenzylamine:</strong> 3,4-Dihydroxybenzylamine is used in the synthesis of poly[3,4-dihydroxybenzylamine] (PDHBA) by oxidative polymerization, exploring its application as a lower homolog of dopamine for potential use in synthetic pathways and materials science (Petran et al., 2023).</li>
<li><strong>Detection of Urinary Free Metanephrines:</strong> Utilizing 3,4-Dihydroxybenzylamine hydrobromide as an internal standard, this research enhances the detection accuracy of urinary free metanephrines for diagnosing pheochromocytomas and paragangliomas, showcasing its importance in clinical diagnostic applications (Wang et al., 2020).</li>
<li><strong>Development of HPLC-ECD Method:</strong> A study developed an HPLC-ECD method using 3,4-Dihydroxybenzylamine as an internal standard for the analysis of vitamin C in plasma, demonstrating the chemical&rsquo;s utility in enhancing analytical methodologies in biochemical research (Clark and Frank, 2016).</li>
<li><strong>Fluorescence Analysis of Catecholamines:</strong> 3,4-Dihydroxybenzylamine is used as an internal standard to determine catecholamines and related compounds in rat brain tissue, underlining its application in neurochemical analysis and research (Fonseca et al., 2017).</li>
</ul>

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E C Chan et al.
Rapid communications in mass spectrometry : RCM, 14(21), 1959-1964 (2000-11-21)
An assay of norepinephrine (NE), epinephrine (E), dopamine (DA), normetanephrine (NE) and metanephrine (MN) based on high-performance liquid chromatography (HPLC) in combination with atmospheric pressure chemical ionization mass spectrometry (APcI-MS) is described. The catecholamines and metanephrines were extracted from urine
J A Prezioso et al.
Pigment cell research, 3(2), 49-54 (1990-03-01)
The rationale for melanoma specific dihydroxybenzene containing antitumor agents is based in part upon the ability of the enzyme tyrosinase to oxidize these pro drugs to toxic intermediates. In situ tyrosinase activity was demonstrated to be affected by both cell
H B He et al.
Journal of chromatography, 574(2), 213-218 (1992-02-14)
This paper describes the application of a window diagram technique to optimize the four components of eluent (sodium acetate, sodium heptanesulfonate, acetonitrile and pH adjusted by monochloroacetic acid), for complete separation of five catecholamine compounds and the internal standard (3,4-dihydroxybenzylamine
F Boomsma et al.
Journal of cardiovascular pharmacology, 22(2), 198-202 (1993-08-01)
We noted rapid breakdown at 4 degrees and 20 degrees C of dopamine (DA) (but not of (nor)epinephrine and epinine) in pig plasma, but not in human plasma. The enzyme responsible appears to be a semicarbazide-sensitive amine oxidase (SSAO) because
J A Prezioso et al.
The Journal of investigative dermatology, 99(3), 289-293 (1992-09-01)
Buthionine sulfoximine (BSO), a specific inhibitor of glutathione synthesis, showed variable growth-inhibitory activity in different tumor cell lines with a high degree of inhibitory activity against melanoma-derived cell lines. A correlation between BSO growth-inhibitory effects and cellular glutathione peroxidase activity

Questions

  1. Can it be stored at -20C

    1 answer
    1. This product is stored at room temperature. In general, powder materials are not effected by freezer storage, however this item has not been tested under these storage conditions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service