Skip to Content
Merck
All Photos(1)

Key Documents

81080

Sigma-Aldrich

Chloroplatinic acid hydrate

~38% Pt basis

Synonym(s):

Hexachloroplatinic(IV) acid hydrate, Hydrogen hexachloroplatinate(IV) hydrate, Platinic chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · xH2O
CAS Number:
Molecular Weight:
409.81 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

powder

reaction suitability

core: platinum
reagent type: catalyst

concentration

~38% Pt

mp

60 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

SMILES string

O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.H2O.Pt/h6*1H;1H2;/q;;;;;;;+4/p-4

InChI key

SLIOYUPLNYLSSR-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Reduction of carboxamides with hydrosilanes
  • Hydrosilylation of siloxanes
  • Dehydrocoupling polycondensation reactions
  • Hydrolysis of trichlorosilanes

Storage and Stability

1. Hygroscopic material, always handle in a glove bag or a glove box filled with dry inert gas
2. Recommended for one time use to prevent water absorption and degradation. If necessary, once opened, repack and seal under dry inert gas.
3. Store with care. Prevent from light. Always store in well-sealed containers in a dry inert atmosphere. Recommended to store containers in a secondary desiccator with desiccant in a dry inert atmosphere.
Packaged under dry Nitrogen gas

Other Notes

Catalyst for the hydrosilylation of olefins

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan R Bassindale et al.
Chemical communications (Cambridge, England), (12), 1382-1383 (2003-07-05)
Hexasilsesquioxane cages (T6) have been prepared from a range of alkyl and aryl trichlorosilanes using a "non aqueous" hydrolysis with dimethyl sulfoxide.
Toal, S. J.; et al.
Organometallics, 24, 3081-3081 (2005)
de Vekki, D. A.; et al.
Russ. J. Gen. Chem., 75, 1757-1757 (2005)
Shiori Hanada et al.
Journal of the American Chemical Society, 131(41), 15032-15040 (2009-09-30)
The synergetic effect of two Si-H groups leads to efficient reduction of carboxamides to amines by platinum catalysts under mild conditions. The rate of the reaction is dependent on the distance of two Si-H groups; 1,1,3,3-tetramethyldisiloxane (TMDS) and 1,2-bis(dimethylsilyl)benzene are
J.L. Speier
Adv. Organomet. Chem., 17, 407-407 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service