Skip to Content
Merck
All Photos(1)

Key Documents

795569

Sigma-Aldrich

Garg 4,5-indolyne precursor

95%

Synonym(s):

4-(Trimethylsilyl)-1H-indol-5-yl trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14F3NO3SSi
CAS Number:
Molecular Weight:
337.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

Quality Level

Assay

95%

form

solid

mp

43-48 °C

functional group

fluoro
triflate

storage temp.

2-8°C

SMILES string

O=S(OC1=C([Si](C)(C)C)C2=C(NC=C2)C=C1)(C(F)(F)F)=O

InChI

1S/C12H14F3NO3SSi/c1-21(2,3)11-8-6-7-16-9(8)4-5-10(11)19-20(17,18)12(13,14)15/h4-7,16H,1-3H3

InChI key

LLCJFYYASDIOJN-UHFFFAOYSA-N

General description

Garg 4,5-indolyne precursor is an indolyne precursor developed by Professor Neil Garg and coworkers. It is widely used for the synthesis of various N-heterocyclic based compounds. It is also useful for the preparation of substituted indoles (Diels–Alder and azide cycloaddition products).

Application

Indolyne precursor can be activated under mild fluoride conditions, which are highly reactive with dienes and other dipolariphiles (e.g. azides) to provide cycloaddition products containing an indole motif.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

Related Content

The Garg group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service