Skip to Content
Merck
All Photos(2)

Key Documents

791326

Sigma-Aldrich

Titanium(IV) oxide, brookite

nanopowder, <100 nm, 99.99% trace metals basis

Synonym(s):

Brookite, Titania

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

nanopowder

particle size

<100 nm

mp

1843 °C (lit.)

Application

Brookite;the orthorhombic polymorph of Titanium(IV) Oxide;has potential in fields including photolysis;and the synthesis of nanowires

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin;Shu;
Journal of Materials Chemistry, 15(6), 674-682 (2005)
Xiang-dong Meng;
Mat. Res. Bul., 39(14-15), 2163-2170 (2004)
Ting Cheng et al.
Acta biomaterialia, 54, 186-200 (2017-03-21)
Near-infrared (NIR) triggered chemical delivery allows on-demand release with the advantage of external tissue stimulation. Bioresorbable polyester poly-l-lactic acid (PLLA) was compounded with photoadditives of neat zinc oxide (ZnO) nanoparticles and 980→365nm LiYF Upconverting nanoparticles and zinc oxide nanorods were

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service