Skip to Content
Merck
All Photos(1)

Key Documents

767654

Sigma-Aldrich

cis-Dichlorobis(dimethyl sulfoxide)platinum(II)

97%

Synonym(s):

cis-Dichlorobis(dimethylsulfoxido)platinum(II), Pt(DMSO)2Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12Cl2O2PtS2
CAS Number:
Molecular Weight:
422.25
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

218-224 °C

SMILES string

Cl[Pt]Cl.CS(C)=O.CS(C)=O

InChI

1S/2C2H6OS.2ClH.Pt/c2*1-4(2)3;;;/h2*1-2H3;2*1H;/q;;;;+2/p-2

InChI key

KDADQDWVKZJTDQ-UHFFFAOYSA-L

General description

cis-Dichlorobis(dimethyl sulfoxide)platinum(II) is a platinum complex used as a catalyst in various chemical reactions, including the methylation of polyfluorinated aryl imines and methane oxidation. It is identified as an effective C-F activation precatalyst.

Application

cis-Dichlorobis(dimethyl sulfoxide)platinum(II) can be used as a:
  • Catalyst in the oleum-mediated (SO3-H2SO4) methane oxidation to form methyl bisulfate (MBS).
  • Precursor in the synthesis of cyclometalated complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gorodetsky et al.
Medical physics, 20(4), 1007-1012 (1993-07-01)
Noninvasive analysis of heavy elements in external tissues by diagnostic-x-ray spectrometry (DXS) is presented. Pt can be detected accurately with sensitivity below 1 microgram/g wet weight of tissue. In the present paper the possibility to monitor Pt accumulation and clearance
K Nishio et al.
International journal of cancer, 55(4), 616-622 (1993-10-21)
cis-Diamminedichloroplatinum(II) (CDDP) induced G2-phase arrest in PC-9 human cancer cells. To elucidate how CDDP acts on cell-cycle regulation, we analyzed the effect of CDDP on cell-cycle regulators such as p34cdc2 protein kinase. p34cdc2 protein kinase activity was maximum in G2
Treatment of pathological stage T3N+M0 bladder cancer: the case for observation.
D A Corral et al.
Seminars in urology, 11(2), 64-68 (1993-05-01)
K R Grigorian et al.
Biofizika, 56(1), 31-34 (2011-03-30)
The interaction of cis-[PtCl2(Me2SO)2] with human serum albumin (HSA) and the sensitivity of the complex towards the thermal denaturation depending on the duration of incubation have been studied by absorption and fluorescence spectroscopy methods. Optimum conditions for cis-[PtCl2(Me2SO)2] binding to
B A Teicher et al.
Cancer chemotherapy and pharmacology, 32(4), 315-319 (1993-01-01)
The survival of bone-marrow granulocyte-macrophage colony-forming units (CFU-GM), an alkylating-agent-sensitive normal tissue, was assessed in mice bearing the EMT-6 parental tumor or the in vivo resistant EMT-6/CDDP, EMT-6/CTX, EMT-6/Thio, and EMT-6/Carbo tumors. The survival pattern of the bone-marrow CFU-GM recapitulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service