Skip to Content
Merck
All Photos(1)

Documents

765090

Sigma-Aldrich

Polyethylenimine, linear

average Mn 10,000, PDI ≤1.3

Synonym(s):

PEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2NH)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000

mp

48-53 °C

PDI

≤1.3

storage temp.

2-8°C

InChI

1S/C2H5N/c1-2-3-1/h3H,1-2H2

InChI key

NOWKCMXCCJGMRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylenimine (linear) is a versatile cationic polymer with a high transfection efficiency. It is used as a gene delivery reagent. It is a non-lipid polycation that can be used to transfect oligonucleotides and plasmid into cells in vitro and in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparison of linear and branched polyethylenimine (PEI) with DCChol/DOPE liposomes for gene delivery to epithelial cells in vitro and in vivo
Wiseman JW, et al.
Gene Therapy, 10(19), 1654-1654 (2003)
Systemic linear polyethylenimine (L-PEI)-mediated gene delivery in the mouse
Zou S, et al.
The journal of gene medicine, 2(2), 128-134 (2000)
Bromodomain and extra-terminal (BET) protein inhibitors suppress chondrocyte differentiation and restrain bone growth
Niu N, et al.
The Journal of biological chemistry, 291(52), 26647-26657 (2016)
Dae Hee Lee et al.
Biomaterials, 171, 34-45 (2018-04-22)
Limitation of current anti-Vascular Endothelial Growth Factor (VEGF) cancer therapy is transitory responses, inevitable relapses and its insufficient tumor-targeting. Thus, multifaceted approaches, including the development of bispecific antibodies and combination strategies targeting different pathways have been proposed as an alternative.
Marie-Elise Bonnet et al.
Pharmaceutical research, 25(12), 2972-2982 (2008-08-19)
The success of nucleic acid therapies depends upon delivery vehicle's ability to selectively and efficiently deliver therapeutic nucleic acids to target organ with minimal toxicity. The cationic polymer polyethylenimine (PEI) has been widely used for nucleic acid delivery due to

Articles

This review will focus on the sub-sector of drug delivery concerning the delivery of nucleic acids, with a particular focus on poly(ethyleneimine).

Gene therapy has become one of the most discussed techniques in biomedical research in recent years.

Professor Yoshiki Katayama (Kyushu University, Japan) discusses recent advances in drug delivery systems and strategies that exploit the EPR effect, with a special focus on stimuli-responsive systems based on novel materials.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service