Skip to Content
Merck
All Photos(1)

Key Documents

764396

Sigma-Aldrich

(E)-Cyclooct-4-enol

Synonym(s):

(4E)-4-Cycloocten-1-ol, trans-Cyclooctenol, TCO-OH, rel-(1R,4E,pR)-Cyclooct-4-enol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

hydroxyl

storage temp.

−20°C

SMILES string

O[C@H]1CC/C=C/CCC1

InChI

1S/C8H14O/c9-8-6-4-2-1-3-5-7-8/h1-2,8-9H,3-7H2/b2-1+/t8-/m0/s1

InChI key

UCPDHOTYYDHPEN-CMLYIYFCSA-N

Application

(E)-Cyclooct-4-enol may be used in the multi-step synthesis of trans-cyclooctene geranyl diphosphate, a novel strained-ring containing protein farnesyltransferase (PFTase) substrate that can be used in site-specific modification technologies.
Hydroxyl modified cyclooctene derivative. Cyclooctenes are useful in strain-promoted copper-free click chemistry cycloaddition reactions with 1,2,4,5-tetrazines. This cyclooctene will react with tetrazine functionalized compounds or biomolecules without the need for a catalyst to result in a stable covalent linkage. The 4+2 inverse electron demand Diels-Alder cycloaddition between trans-cyclooctene and tetrazines is the fastest biologically compatible ligation technology reported and has had many applications in biological labeling and imaging.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reprint of: Synthesis, characterization and radiolabeling of carborane-functionalized tetrazines for use in inverse electron demand Diels?Alder ligation reactions.
Genady AR, et al.
Journal of Organometallic Chemistry, 798, 278-288 (2015)
Development of a novel antibody?tetrazine conjugate for bioorthogonal pretargeting.
Maggi A, et al.
Organic & Biomolecular Chemistry, 14(31), 7544-7551 (2016)
A 99mTc-Labelled Tetrazine for Bioorthogonal Chemistry. Synthesis and Biodistribution Studies with Small Molecule trans-Cyclooctene Derivatives.
Vito A, et al.
PLoS ONE, 11(12), e0167425-e0167425 (2016)
Click to release: instantaneous doxorubicin elimination upon tetrazine ligation.
Versteegen RM, et al.
Angewandte Chemie (International Edition in English), 52(52), 14112-14116 (2013)
Site?Specific Labeling of Proteins and Peptides with Trans?cyclooctene Containing Handles Capable of Tetrazine Ligation
Wollack JW, et al.
Chemical Biology & Drug Design, 84(2), 140-147 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service