Skip to Content
Merck
All Photos(1)

Key Documents

762385

Sigma-Aldrich

6,6′-Bis(4-(S)-isopropyl-2-oxazolinyl)-2,2′-bipyridine

96%

Synonym(s):

Bipymox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N4O2
CAS Number:
Molecular Weight:
378.47
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]/D -97.0°, c = 0.5% in dichloromethane

reaction suitability

reaction type: Asymmetric synthesis
reagent type: ligand
reaction type: Hydrosilylations

mp

185-190 °C

SMILES string

CC(C)[C@H]1COC(=N1)c2cccc(n2)-c3cccc(n3)C4=N[C@H](CO4)C(C)C

InChI

1S/C22H26N4O2/c1-13(2)19-11-27-21(25-19)17-9-5-7-15(23-17)16-8-6-10-18(24-16)22-26-20(12-28-22)14(3)4/h5-10,13-14,19-20H,11-12H2,1-4H3/t19-,20-/m1/s1

InChI key

NNCROLGXDLXLJM-WOJBJXKFSA-N

Application

Ligand used in the asymmetric hydrosilation of ketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishiyama, H.; Yamaguchi, S. et al.
Tetrahedron Asymmetry, 4, 143-143 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service