Skip to Content
Merck
All Photos(7)

Key Documents

761435

Sigma-Aldrich

cataCXium® A Pd G3

greener alternative

95%

Synonym(s):

Mesylate[(di(1-adamantyl)-n-butylphosphine)-2-(2′-amino-1,1′-biphenyl)]palladium(II), [(Di(1-adamantyl)-butylphosphine)-2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, cataCXium-A-Pd-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H52NO3PPdS
CAS Number:
Molecular Weight:
728.27
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

greener alternative product score

old score: 16
new score: 2
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤3% acetone

mp

196-241 °C (decomposition)

functional group

phosphine

greener alternative category

SMILES string

CS(=O)(=O)O[Pd]c1ccccc1-c2ccccc2N.CCCCP([C@@]34C[C@@H]5C[C@@H](C[C@@H](C5)C3)C4)[C@@]67C[C@@H]8C[C@@H](C[C@@H](C8)C6)C7

InChI

1S/C24H39P.C12H10N.CH4O3S.Pd/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h17-22H,2-16H2,1H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1/t17-,18+,19-,20-,21+,22-,23-,24-;;;

InChI key

REYVZCOGMIXVNX-DVBMAMJVSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Atom Economy”. Click here to view its DOZN scorecard.

Application

cataCXium® A Pd G3 is a Buchwald third-generation precatalyst that can be used in:
  • Direct ortho-arylation of pyridinecarboxylic acids.[1][2]
  • Catalyzing Suzuki–Miyaura cross-coupling in the synthesis of 1-heteroaryl-3-azabicyclo[3.1.0]hexanes.[3]
  • Palladium-catalyzed carbonylative carboperfluoroalkylation of alkynes.[4]
  • Suzuki–Miyaura coupling reaction of geminal bis(boryl)cyclopropanes in the synthesis of various gem-disubstituted cyclopropanes.[5]
  • Boroperfluoroalkylation of terminal alkynes.[6]
  • Copper-free Sonogashira coupling reaction of aromatic halides with alkynes to form C-C bond.[7]
  • Suzuki cross-coupling between organotrifluoroborate and aryl halides.[8]
For small scale and high throughput uses, product is also available as ChemBeads (928305)

Legal Information

cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-free Sonogashira cross-coupling reactions: an overview
Mohajer F, et al.
Royal Society of Chemistry Advances, 11, 6885-6925 (2021)
Construction of 1-Heteroaryl-3-azabicyclo [3.1. 0] hexanes by sp3-sp2 Suzuki-Miyaura and Chan-Evans-Lam Coupling Reactions of Tertiary Trifluoroborates.
Harris MR, et al.
Organic Letters, 19(9), 2450-2453 (2017)
A Modular Approach to the Synthesis of gem-Disubstituted Cyclopropanes.
Harris MR, et al.
Organic Letters, 20(10), 2867-2871 (2018)
The use of carboxylic acids as traceless directing groups for regioselective C-H bond functionalisation.
Font M and Quibell
Chemical Communications (Cambridge, England), 53(41), 5584-5597 (2017)
Rapid access to diverse, trifluoromethyl-substituted alkenes using complementary strategies
Phelan J, et al.
Chemical Science, 9, 3215-3220 (2018)

Articles

All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service