Skip to Content
Merck
All Photos(1)

Key Documents

753629

Sigma-Aldrich

Gold nanoparticles

30 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

concentration

~1.8E+11 particles/mL

OD

1

diameter

30 nm

λmax

523-526 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles (AuNPs), 30 nm are quasi dynamic colloidal nanomaterials with good surface chemistry and high surface to volume ratio. Their properties include high stability, biocompatibility, and low toxicity which make them useful in biological applications.

Application

AuNPs can be used for a variety of applications such as biomedical, electrochemical and energy based devices. They can also be used in drug delivery and biosensing based device fabrication.

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold Nanoparticles For Physics, Chemistry And Biology (Second Edition) (2012)
Akanksha Raj et al.
Toxicology and industrial health, 36(10), 769-778 (2020-11-27)
With the extensive usage of gold nanoparticles (AuNPs) in various industrial sectors and biomedical applications, evaluation of their possible effects on human health becomes imperative. Therefore, the present study was aimed toward assessing the dose-dependent impact of AuNPs ingestion on
Graphene-gold nanoparticles hybrid?synthesis, functionalization, and application in a electrochemical and surface-enhanced raman scattering biosensor
Khalil I, et al.
Materials, 9(6), 406-406 (2016)
Gold nanoparticles for applications in cancer radiotherapy: Mechanisms and recent advancements
Her S, et al.
Advanced Drug Delivery Reviews, 109(6), 84-101 (2017)
Spectroscopic Study of the Interaction of Carboxyl-Modified Gold Nanoparticles with Liposomes of Different Chain Lengths and Controlled Drug Release by Layer-by-Layer Technology
Kanwa N, et al.
The Journal of Physical Chemistry B, 121(50), 11333-11343 (2017)

Articles

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service