Skip to Content
Merck
All Photos(2)

Documents

752207

Sigma-Aldrich

Biphenyl-4-thiol

97%

Synonym(s):

1,1′-Biphenyl-4-thiol, 4-Biphenylylthiol, 4-Mercaptobiphenyl, 4-Phenylbenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10S
CAS Number:
Molecular Weight:
186.27
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

108-112 °C

storage temp.

2-8°C

SMILES string

Sc1ccc(cc1)-c2ccccc2

InChI

1S/C12H10S/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13H

InChI key

KRVHFFQFZQSNLB-UHFFFAOYSA-N

General description

Biphenyl-4-thiol(BPT) is an alkylthiol that forms a self-assembled monolayer(SAM) which can be used as a spacer chain that enhances the adhesion and frictional forces on a variety of surfaces.

Application

BPT may also be used as a resist in electron lithography to form lubricants for microelectromechanical systems(MEMS). BPT may be used to surface modify gold(Au) which can be potentially used in the formation of freestanding nanosheets.
This material is used to create close-packed monolayers in large-area molecular junctions.† It also finds use as a negative resist material for nanolithography.†

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Freestanding nanosheets from crosslinked biphenyl self-assembled monolayers.
Eck W, et al.
Advanced Materials, 17(21), 2583-2587 (2005)
Kronemeijer; A. J.; Huisman; E. H.; Akkerman; H. B.; et al
Applied Physics Letters, 97, 173302/1-173302/3 (2010)
Hyunook Kang et al.
Talanta, 168, 240-245 (2017-04-11)
We report a method of small molecule analysis using a converted graphene-like monolayer (CGM) plate and laser desorption/ionization time-of-flight mass spectrometry (LDI-TOF MS) without organic matrices. The CGM plate was prepared from self-assembled monolayers of biphenyl-4-thiol on gold using electron
Joong Bum Lee et al.
ACS nano (2020-12-04)
Deterministic positioning and assembly of colloidal nanoparticles (NPs) onto substrates is a core requirement and a promising alternative to top-down lithography to create functional nanostructures and nanodevices with intriguing optical, electrical, and catalytic features. Capillary-assisted particle assembly (CAPA) has emerged
Giovanni Luca Bartolomeo et al.
Applied spectroscopy, 74(11), 1358-1364 (2020-03-14)
Tip-enhanced Raman spectroscopy (TERS) is a powerful tool for the characterization of surfaces and two-dimensional materials, delivering both topographical and chemical information with nanometer-scale spatial resolution. Atomic force microscopy (AFM)-TERS combines AFM with a Raman spectrometer and is a very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service