Skip to Content
Merck
All Photos(1)

Key Documents

749613

Sigma-Aldrich

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate

97%

Synonym(s):

DMTMM, MMTM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17BF4N4O3
Molecular Weight:
328.07
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

202-207 °C

functional group

ether

SMILES string

F[B-](F)(F)F.COc1nc(OC)nc(n1)[N+]2(C)CCOCC2

InChI

1S/C10H17N4O3.BF4/c1-14(4-6-17-7-5-14)8-11-9(15-2)13-10(12-8)16-3;2-1(3,4)5/h4-7H2,1-3H3;/q+1;-1

InChI key

LCRXDNPNQMIQFZ-UHFFFAOYSA-N

Application

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate (DMTMM BF4) can be used as a coupling reagent alternative to DMTMM Cl in peptide synthesis because of its stability. It is also used in the synthesis of:
  • Esters and peptides in both solution and solid-phase.
  • γ-aminobutyric acid (GABA)-containing cyclic heptapeptide, unguisin A.
  • The cyclization of linear tetrapeptides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Laura Alfieri et al.
Antioxidants (Basel, Switzerland), 9(3) (2020-03-22)
The ability of gelatin-based hydrogels of incorporating and releasing under controlled conditions 5,6-dihydroxyindole-2-carboxylic acid (DHICA), a melanin-related metabolite endowed with marked antioxidant properties was investigated. The methyl ester of DHICA, MeDHICA, was also tested in view of its higher stability
Diego Serrano et al.
Molecular cell, 80(6), 1025-1038 (2020-12-11)
The structural organization of chromosomes is a crucial feature that defines the functional state of genes and genomes. The extent of structural changes experienced by genomes of eukaryotic cells can be dramatic and spans several orders of magnitude. At the
An update on new methods to synthesize cyclotetrapeptides
Rodriguez LML, et al
Organic & Biomolecular Chemistry, 13(25), 6906-6921 (2015)
Dongmei Zhang et al.
ACS applied materials & interfaces, 12(5), 5542-5556 (2020-01-16)
An efficient and available material for promoting skin regeneration is of great importance for public health, but it remains an elusive goal. Inspired by fetal scarless wound healing, we develop a wearable biomimetic film (WBMF) composed of hyaluronan (HA), vitamin
Felix Klatt et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(6), 2894-2905 (2020-01-29)
The Mediator kinase module regulates eukaryotic transcription by phosphorylating transcription-related targets and by modulating the association of Mediator and RNA polymerase II. The activity of its catalytic core, cyclin-dependent kinase 8 (CDK8), is controlled by Cyclin C and regulatory subunit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service