Skip to Content
Merck
All Photos(1)

Documents

747068

Sigma-Aldrich

Poly(N-isopropylacrylamide), azide terminated

average Mn 15,000, PDI ≤1.4

Synonym(s):

Azide PolyNIPAM, PNIPAAm, PNIPAM-N3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn 15,000

PDI

≤1.4

storage temp.

2-8°C

Application

Poly(N-isopropylacrylamide) is a stimuli responsive polymer and synthesized by controlled radical polymerization to achieve a narrow polydispersity. The azide functionality is commonly used as a click group which can react with acetylene/alkyne groups. This polymer can be used to bind to proteins to form stable protein-polymer hybrids with responsive properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecular Rapid Communications, 29, 1172-1176 (2008)

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service