Skip to Content
Merck
All Photos(1)

Key Documents

73328

Sigma-Aldrich

Trimethylsilyl cyanide

technical, ≥95% (GC)

Synonym(s):

Cyanotrimethylsilane, TMSCN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCN
CAS Number:
Molecular Weight:
99.21
Beilstein:
1737612
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥95% (GC)

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

114-117 °C (lit.)

mp

8-11 °C (lit.)

density

0.793 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)C#N

InChI

1S/C4H9NSi/c1-6(2,3)4-5/h1-3H3

InChI key

LEIMLDGFXIOXMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trimethylsilyl cyanide (TMSCN) can be used as a reagent in the:
  • Cyanosilylation of carbonyl compounds using various catalysts.
  • Synthesis of α-aminonitriles by one-pot, three-component Strecker reaction of ketones with various amines using Brønsted acid catalyst.
  • Cyanation of aryl halides using palladium-complex as a catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric addition of trimethylsilyl cyanide to ketones catalyzed by Al (salen)/triphenylphosphine oxide
Kim SS and Kwak Ju M
Tetrahedron, 62(1), 49-53 (2006)
Indium tribromide: a highly effective catalyst for the addition of trimethylsilyl cyanide to ?-hetero-substituted ketones
Bandini M, et al.
Tetrahedron Letters, 42(16), 3041-3043 (2001)
A convenient and efficient procedure for the palladium-catalyzed cyanation of aryl halides using trimethylsilylcyanide
Sundermeier M, et al.
Journal of Organometallic Chemistry, 684(1-2), 50-55 (2003)
Zhiguo Zhang et al.
The Journal of organic chemistry, 76(23), 9764-9776 (2011-10-21)
We report a new thiourea-Brønsted acid cooperative catalytic system for the enantioselective cyanosilylation of aldehydes with yields up to 90% and enantioselectivities up to 88%. The addition of an achiral acid was found to be crucial for high asymmetric induction.
Gang Chen et al.
Organic letters, 10(20), 4573-4576 (2008-09-24)
A convenient and efficient synthesis of alpha-aryl nitriles was developed by direct cyanation of alcohols with TMSCN under the catalysis of Lewis acid. Using 5-10 mol % of InBr3 as the catalyst, a variety of benzylic alcohols can be converted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service