Skip to Content
Merck
All Photos(1)

Documents

73261

Sigma-Aldrich

1-Butyl-4-methylpyridinium tetrafluoroborate

≥97.0% (T)

Synonym(s):

1-Butyl-4-picolinium tetrafluoroborate, 4MBPBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16BF4N
CAS Number:
Molecular Weight:
237.05
Beilstein:
9019838
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

form

liquid

refractive index

n20/D 1.4

density

1.20 g/mL at 20 °C (lit.)

SMILES string

F[B-](F)(F)F.CCCC[n+]1ccc(C)cc1

InChI

1S/C10H16N.BF4/c1-3-4-7-11-8-5-10(2)6-9-11;2-1(3,4)5/h5-6,8-9H,3-4,7H2,1-2H3;/q+1;-1

InChI key

VISYYHYJMCAKAF-UHFFFAOYSA-N

General description

Pyridinium based ionic liquid

Application

4MBPBF4 has been used in the preparation of the SWNT-polymer composite films to improve the dispersion of SWNTs and conductivity of the composites (SWNT= singlewalled carbon nanotubes). The use of 4MBPBF4 as a reaction media during derivatization of dimethyl sulfate with dibenzazepine, accelerates the rate of the reaction. It can also be used to modify carbon paste electrode, which leads to high sensitivity, selectivity and low detection limit for both potassium ferricyanide and dopamine by cyclic voltammetric technique.

Other Notes

Lipase catalyzed kinetic resolution in ionic liquids with improved enantioselectivity

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Transparent Stretchable Single-Walled Carbon Nanotube-Polymer Composite Films with Near-Infrared Fluorescence.
Ma R, et al.
Advanced Materials, 25(18), 2548-2553 (2013)
S. Schofer et al.
Chemical Communications (Cambridge, England), 425-425 (2001)
Electrochemical investigations of potassium ferricyanide and dopamine by 1-butyl-4-methylpyridinium tetrafluoro borate modified carbon paste electrode: A cyclic voltammetric study.
Pandurangachar M, et al.
International Journal of Electrochemical Science, 5(8), 1187-1202 (2010)
Mixed ionic liquid as electrolyte for lithium batteries
Diaw, M., et al.
Journal of Power Sources, 146.1, 682-684 (2005)
Mixing thermodynamic properties of 1-butyl-4-methylpyridinium tetrafluoroborate [b4mpy][BF 4] with water and with an alkan-1ol (methanol to pentanol)
Ortega, J., et al.
The Journal of Chemical Thermodynamics, 40.7, 1087-1094 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service