Skip to Content
Merck
All Photos(1)

Key Documents

730637

Sigma-Aldrich

Dihydrotestosterone-2,3,4-13C3 solution

0.1 mg/mL in methanol, 99 atom % 13C, 97% (CP)

Synonym(s):

5α−Androstan-17β-ol-3-one-13C3, Androstanolone-13C3, Stanolone-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C16H30O2
Molecular Weight:
293.42
EC Number:
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

solution

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

concentration

0.1 mg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C
−70°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CC[C@H]4[13CH2][13C](=O)[13CH2]C[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1/i7+1,11+1,13+1

InChI key

NVKAWKQGWWIWPM-PUXGIKSBSA-N

Related Categories

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renaud Mevel et al.
eLife, 9 (2020-10-08)
The characterization of prostate epithelial hierarchy and lineage heterogeneity is critical to understand its regenerative properties and malignancies. Here, we report that the transcription factor RUNX1 marks a specific subpopulation of proximal luminal cells (PLCs), enriched in the periurethral region

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service