Skip to Content
Merck
All Photos(2)

Key Documents

724254

Sigma-Aldrich

1,4-Benzodioxan-6-sulfonyl chloride

95%

Synonym(s):

2,3-Dihydro-1,4-benzodioxine-6-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO4S
CAS Number:
Molecular Weight:
234.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

65-70 °C

SMILES string

ClS(=O)(=O)c1ccc2OCCOc2c1

InChI

1S/C8H7ClO4S/c9-14(10,11)6-1-2-7-8(5-6)13-4-3-12-7/h1-2,5H,3-4H2

InChI key

JWHSRWUHRLYAPM-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service