Skip to Content
Merck
All Photos(1)

Key Documents

721301

Sigma-Aldrich

Sodium pregnenolone-17α,21,21,21-d4 sulfate

98 atom % D, 98% (CP)

Synonym(s):

3β-Hydroxy-5-pregnen-20-one-17α,21,21,21-d4 3-sulfate, 3-Hydroxypregn-5-en-10-one-17α,21,21,21-d4 sulfate, 5-Pregnen-3β-ol-20-one-17α,21,21,21-d4 sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D4H27NaO5S
Molecular Weight:
422.55
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

192 °C (lit.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

[Na+].[2H]C([2H])([2H])C(=O)[C@]1([2H])CC[C@H]2[C@@H]3CC=C4CC(CC[C@]4(C)C3CC[C@]12C)OS([O-])(=O)=O

InChI

1S/C21H32O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,15-19H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t15?,16-,17-,18-,19?,20-,21+;/m0./s1/i1D3,17D;

InChI key

QQVJEIZJHDPTSH-KSVJGDSJSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service