Skip to Content
Merck
All Photos(1)

Documents

710806

Sigma-Aldrich

18-Hydroxycorticosterone

97% (CP)

Synonym(s):

4-Pregnene-11β,18,21-triol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein:
2631066
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(CO)[C@@]3([H])CC[C@]4([H])C(CO)=O)=C1

InChI

1S/C21H30O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,14-17,19,22-23,25H,2-7,9-11H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1

InChI key

HFSXHZZDNDGLQN-ZVIOFETBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
To best serve customer requests, each order is packaged and shipped on demand approximately 2 weeks after receipt of an order. If you have questions regarding availability and price, please contact the Stable Isotope Customer Service Group at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at isosales@sial.com.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Honda et al.
Endocrinologia japonica, 29(5), 529-540 (1982-10-01)
In order to systematically analyze the regulation and metabolism of steroid hormones in a case of primary aldosteronism with multiple lesions, including adenoma and nodular hyperplasia of the left adrenal gland, the amounts of 9 steroids (progesterone (P), 11-deoxycorticosterone (DOC)
J Palmer et al.
Electrophoresis, 19(16-17), 3045-3051 (1998-12-31)
Separation of endogenous 17- or 18-hydroxylated corticosteroids (of the 21-hydroxylated 4-pregnen series) as charged chelates in capillary electrophoresis with borate as the ligand is demonstrated. Aldosterone, 18-hydroxycorticosterone, 18-hydroxy-11-deoxycorticosterone, cortisone, cortisol, and 11-deoxycortisol are separated and resolved by 400 mM borate
M K Birmingham et al.
Endocrine research, 24(3-4), 595-599 (1999-01-15)
A simple semantic differential test was administered twice daily and urinary hormones measured as described previously . The subjects included the patient and two controls, one of whom experienced premenstrual physical, but not mental, discomfort. The test scores of the
P Vecsei et al.
Clinical chemistry, 28(3), 453-456 (1982-03-01)
For distinguishing primary aldosteronism from essential hypertension, we use simple direct radioimmunoassays for "aldosterone" (aldosterone and other materials that react with the antibody to aldosterone) and "18-hydroxycorticosterone" (similarly) in unprocessed urine. Patients with primary aldosteronism have high values for "aldosterone."
M Peter et al.
The Journal of clinical endocrinology and metabolism, 80(5), 1622-1627 (1995-05-01)
Aldosterone (Aldo), the most potent mineralocorticoid, is synthesized in the adrenal zona glomerulosa, requiring 11 beta-hydroxylation of 11-deoxycorticosterone (DOC) to form corticosterone (B), hydroxylation at position C18 to form 18-hydroxycorticosterone (18-OHB), and finally oxidation at position C18. There is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service