Skip to Content
Merck
All Photos(2)

Documents

705039

Sigma-Aldrich

2-Morpholinopyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13BN2O3
CAS Number:
Molecular Weight:
208.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

90-94 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cccnc1N2CCOCC2

InChI

1S/C9H13BN2O3/c13-10(14)8-2-1-3-11-9(8)12-4-6-15-7-5-12/h1-3,13-14H,4-7H2

InChI key

SWDPLAALYQOCHB-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service