Skip to Content
Merck
All Photos(2)

Key Documents

704261

Sigma-Aldrich

Lanthanum strontium manganite

greener alternative

LSM-35

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

powder

mol wt

Mw 223.89 g/mol

composition

La0.65Sr0.35MnO3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

4-8 m2/g

particle size

0.6-1.2 μm (Horiba-Laser)

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

LSM is one of the materials that is useful as an electrode material in solid oxide fuel cells.
Lanthanum strontium manganite (LSM-35) is a ferromagnetic material, a Sr-doped LaMnO3 that is mainly used as a cathode material for solid oxide fuel cells (SOFCs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous slurry processing of monolithic films for SOFC-YSZ, LSM and YSZ-NiO systems
Ramanathan S and Kakade MB
International Journal of Hydrogen Energy, 36(22), 14956-14962 (2011)
Development of lanthanum strontium manganite perovskite cathode materials of solid oxide fuel cells: a review
Jiang SP
Chemosphere, 43, 6799-6833 (2008)
Alexander H Bork et al.
ACS applied materials & interfaces, 12(29), 32622-32632 (2020-06-20)
Increasing the capacity and kinetics of oxygen exchange in solid oxides is important to improve the performance of numerous energy-related materials, especially those for the solar-to-fuel technology. Dual-phase metal oxide composites of La0.65Sr0.35MnO3-x%CeO2, with x = 0, 5, 10, 20
Synthesis and characterization of lanthanum strontium manganite
Ghosh A, et al.
Scr. Mater., 52(12), 1305-1309 (2005)
Development of lanthanum strontium manganite perovskite cathode materials of solid oxide fuel cells: a review.
Jiang SP
J. Mater. Sci., 43(21), 6799-6833 (2008)

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service