Skip to Content
Merck
All Photos(1)

Key Documents

701874

Sigma-Aldrich

1,3-Bis(N-carbazolyl)benzene

97%

Synonym(s):

1,3-Di(9H-carbazol-9-yl)benzene, 9,9′-(1,3-Phenylene)bis-9H-carbazole, N,N′-Dicarbazolyl-3,5-benzene, mCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H20N2
CAS Number:
Molecular Weight:
408.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

173-178 °C

λmax

292, 338 nm

fluorescence

λem 345, 360 nm in THF

Orbital energy

HOMO 6.1 eV 
LUMO 2.4 eV 

OLED Device Performance

ITO/NPD/mCP/BCPO:FIr6 (8%)/TAZ/LiF/Al

  • Color: blue
  • Max. Luminance: 28718 Cd/m2
  • Max. EQE: 19.8 %
  • Turn-On Voltage: 3 V

ITO/NPD/mCP/BCPO:FIrpic (8%)/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 35677 Cd/m2
  • Max. EQE: 23.5 %
  • Turn-On Voltage: 2.8 V

SMILES string

c1cc(cc(c1)-n2c3ccccc3c4ccccc24)-n5c6ccccc6c7ccccc57

InChI

1S/C30H20N2/c1-5-16-27-23(12-1)24-13-2-6-17-28(24)31(27)21-10-9-11-22(20-21)32-29-18-7-3-14-25(29)26-15-4-8-19-30(26)32/h1-20H

InChI key

MZYDBGLUVPLRKR-UHFFFAOYSA-N

Application

Material for use as a phosphorescent host material

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellie Hudson-Heck et al.
Analytica chimica acta, 1090, 91-99 (2019-10-28)
Thymol blue (TB) is one of a suite of indicator dyes appropriate for spectrophotometric determinations of the pH of aqueous solutions. For measurements of seawater pH, meta-cresol purple (mCP) is most often used, but TB is especially well suited for
Yue Zhou et al.
Bioresource technology, 292, 121951-121951 (2019-08-11)
To create more active adsorption sites on biochar, the Fe3O4/GO/citrus peel-derived magnetic bio-nanocomposite (mGOCP) with hierarchically porous architectures was synthesized by a facile one-pot hydrothermal approach for efficient removal of fluoroquinolone antibiotics ciprofloxacin (CIP) and sparfloxacin (SPA). The characterization analysis
Marek Kirs et al.
Applied and environmental microbiology, 82(22), 6757-6767 (2016-10-30)
Identification of sources of fecal contaminants is needed to (i) determine the health risk associated with recreational water use and (ii) implement appropriate management practices to mitigate this risk and protect the environment. This study evaluated human-associated Bacteroides spp. (HF183TaqMan)
Yafei Wang et al.
Chemistry, an Asian journal, 11(18), 2555-2563 (2016-07-22)
Pi-conjugated organic molecules featuring the donor-bridge-acceptor (D-π-A) structure have been widely used in semiconducting materials owing to their rigid structure, good thermal stability, excellent charge transfer, and high emission efficiency. To investigate the effect of the D-π-A molecular structure on
Brigita Vigante et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(13), 3325-3336 (2018-12-12)
With the aim of developing all-organic bipolar semiconductors with high charge mobility and efficient E-type fluorescence (so-called TADF) as environmentally friendly light-emitting materials for optoelectronic applications, four noble metals-free dyes with linear and V-shapes were designed using accepting pyridine-3,5-dicarbonitrile and

Articles

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Since their discovery, organic light emitting devices (OLEDs) have evolved from a scientific curiosity into a technology with applications in flat panel displays and the potential to revolutionize the lighting market. During their relatively short history, the technology has rapidly advanced, and device efficiencies have increased more than 20-fold, approaching the theoretical limit for internal quantum efficiencies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service