70090
Ethyl myristate
99% (GC)
Synonym(s):
Ethyl tetradecanoate, Myristic acid ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)12COOC2H5
CAS Number:
Molecular Weight:
256.42
Beilstein:
1776382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99% (GC)
form
liquid
refractive index
n20/D 1.436 (lit.)
bp
178-180 °C/12 mmHg (lit.)
mp
11-12 °C (lit.)
density
0.86 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
CCCCCCCCCCCCCC(=O)OCC
InChI
1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3
InChI key
MMKRHZKQPFCLLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Woo-kyoung Lee et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(3), 115-123 (2002-12-07)
An oil-in-water solvent evaporation method was used to prepare cyclosporin A (CyA)-loaded particles varying in size (nanoparticles, 'small-sized' microparticles, 'large-sized' microparticles), polymer compositions [poly(D,L-lactide-co-glycolic acid) (PLGA) 50/50, PLGA 85/15, poly(D,L-lactic acid) (PLA)] and additive fatty acid ester (ethyl myristate; EM).
Sven Hartwig et al.
Forensic science international, 131(2-3), 90-97 (2003-02-19)
Fatty acid ethyl esters (FAEE) can be used as alcohol markers in hair. It was investigated in this study whether this diagnostic method is disturbed by hair care and hair cosmetics. Traces of ethyl myristate, ethyl palmitate, ethyl oleate and
Nadia De Giovanni et al.
Journal of analytical toxicology, 32(2), 156-159 (2008-03-13)
The diagnosis of alcoholism is a topical subject of discussion; in fact, many studies have been published on the determination of biochemical markers useful to this target. Fatty acid ethyl esters (FAEE) are minor metabolites of ethanol, and their usefulness
E Akaho et al.
Journal of pharmaceutical sciences, 70(11), 1225-1228 (1981-11-01)
Interactions of phenolic compounds 4-hexylresorcinol and 3,4-dimethylphenol with esters were studied using hexane-ester cosolvent systems by both phase solubility and partitioning methods. The data obtained by the phase solubility method were variable and could not be analyzed by any mathematical
Neonatal hair analysis as a biomarker for in utero alcohol exposure.
Julia Klein et al.
The New England journal of medicine, 347(25), 2086-2086 (2002-12-20)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service