Skip to Content
Merck
All Photos(3)

Key Documents

700193

Sigma-Aldrich

Polyetherimide

melt index 9 g/10 min (337 °C/6.6kg)

Synonym(s):

Poly(bisphenol A-co-4-nitrophthalic anhydride-co-1,3-phenylenediamine), PEI, Poly(bisphenol A anhydride-co-1,3-phenylenediamine), Poly(bisphenol A dianhydride-co-1,3-phenylenediamine), Poly(ether imide), Poly-[2,2′bis(4-(3,4-dicarboxyphenoxy)phenylpropane)-1,3-phenylene bisimide], Polyetherimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C39H30N2O6)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

melt index

9 g/10 min (337 °C/6.6kg)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

Nc7cc(ccc7)N.O1C(=O)c2c(ccc(c2)Oc3ccc(cc3)C(C)(C)c4ccc(cc4)Oc5cc6c(cc5)C(=O)OC6=O)C1=O

InChI

1S/C31H20O8.C6H8N2/c1-31(2,17-3-7-19(8-4-17)36-21-11-13-23-25(15-21)29(34)38-27(23)32)18-5-9-20(10-6-18)37-22-12-14-24-26(16-22)30(35)39-28(24)33;7-5-2-1-3-6(8)4-5/h3-16H,1-2H3;1-4H,7-8H2

InChI key

VYLFLJXGGIUQKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyetherimide (PEI) is a thermoplastic polymer, containing cyclic amide. PEI is a polyimide condensation polymer derived from the bifunctional carboxylic anhydrides and primary diamines.

Application

Polymer blends of PEI and polyethylene glycol (PEG) have been used to prepare carbon molecular sieves. PEI has been used as matrix in the fabrication of various composites:
  • PEI/ BaTi4O9 composites,
  • PEI/glass fiber composites,
  • Nanocomposite PEI/TiO2-sulfonated polymer membranes,
  • Nanoparticles coated PEI-polyurethane (SiO2/PEI-PU) nanofibrous composite membranes,
  • Single walled nanotubes (SWNTs)/PEI nanocomposites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urakawa O
Encylopedia of Polymeric Nanomaterials, 1-10 (2014)
Lukka Thuyavan Y, et al.
Desalination, 365, 355-364 (2015)
Fabrication of hierarchical structured SiO2/polyetherimide-polyurethane nanofibrous separators with high performance for lithium ion batteries
Zhai Y, et al.
Electrochimica Acta, 154(1), 219-226 (2015)
Effects of hydrothermal aging on glass?fiber/polyetherimide (PEI) composites.
Yilmaz T and Sinmazcelik T
J. Mater. Sci., 45(2), 399-404 (2010)
Effects of BaTi4O9 Concentration on Dielectric Constants of the Polyetherimide/BaTi4O9 Composites
Chia Wei Kuo, et al.
Intelligent Technologies and Engineering Systems, 941-948 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service